Supertek® Wheatstone Bridge
Wheatstone Bridge, 109×10 cm (wooden base)
Catalog # 470149-160
Supplier: Shiv Dial Sud & Sons
Fournisseur #: PH50410
Jump to:
Product Details & Documents
The Wheatstone Bridge is complete with binding posts.
- One Length of AWG 30 Nickel Silver Wire
- Scale Graduated in mm
- Slide Knife-Edge Contact
- Binding Posts
The apparatus consists of one length of AWG 30 nickel silver wire mounted on a scale graduated in mm. The scale features a slide knife-edge contact.
For known and unknown resistance, two strips of heavy aluminum are mounted in each of the two arms of the bridge.
Spécifications
- Désignation:Wheatstone Bridge
- Dimensions:109×10 cm (wooden base)