Ward's® What Influences Enzyme Activity? Kit | Avantor
Ward's® What Influences Enzyme Activity? Kit
Supplier:   Avantor
Ward's® What Influences Enzyme Activity? Kit
Supplier:  Avantor
Certificates
Ward's® What Influences Enzyme Activity? Kit
Ward's® What Influences Enzyme Activity? Kit
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This kit demonstrates the process of starch digestion.
  • Lab Teaches Students to Recognize Breakdown of Starch into Glucose
  • Four Activities in One Kit
  • Pre and Post-Laboratory Assessments Included
  • Estimated Lab Time: 30 to 40 minutes
  • Material for 15 Setups
  • Aligned to NGSS standards (MS/HS: PS1, PS2, LS1)
This lab uses four lab activities and the enzyme diastase of malt to explore the breakdown of starch into glucose. Students perform a starch test and observe the breakdown of starch into glucose to learn how concentration, temperature, and pH affect the overall reaction rate.
The kit contains a teacher’s guide, a student study guide, and material for 15 setups.
Ordering information: This kit includes starch solution, diastase, sodium hydroxide, hydrochloric acid, starch-indicator solution, glass vials, plastic pipettes, test tubes with caps, glucose test strips, sample cups, and spot plates.
Product Family Options
Informations sur le produit
  • Désignation
    No. of Lab Groups
    Availability
    Price
  • What influences enzyme activity? kit
    15
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 1
    Kit of 1 - $226.70
    $226.70
    Kit of 1
       $226.70
  • What influences enzyme activity? refill kit
    15
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 1
    Kit of 1 - $127.00
    $127.00
    Kit of 1
       $127.00