ContactUsLinkComponent
Western Blot Analysis Kit, Edvotek
Catalog # 470120-900
Supplier:  Edvotek
undefined
Western Blot Analysis Kit, Edvotek
Catalog # 470120-900
Supplier:  Edvotek
Supplier Number:  317
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Western blot analysis kit
  • No. of Lab Groups:
    6
  • Cat. no.:
    470120-900
  • Supplier No.:
    317
Specifications
About this item
Protein identification is based on both antibody reactions and antigens in this Western Blot activity.
  • Materials for Six Blots Included
  • Instructions with Background Information
  • Students Research Protein Identification
A denaturing SDS polyacrylamide gel separates the proteins, which are transferred or blotted to a nylon membrane. The membrane is then exposed sequentially to solutions containing the primary antibody, followed by a secondary antibody that is coupled to an enzyme.
The membrane is soaked in a substrate solution to develop the color reaction, which results in identifying the antigen as a band, and the molecular weights of the visible bands are measured using pre-stained protein markers.
Ordering information: This kit contains negative lyophilized control BSA, high and low concentration (lyophilized); an anti-BSA protein antibody; secondary antibody conjugate; hydrogen peroxide; peroxide co-substrate, 10X; blocking buffer; PBS buffer; pre-stained protein standard marker (lyophilized); tris-glycine-SDS electrophoresis buffer; powdered milk; and instructions with background information. The kit requires but does not include three polyacrylamide gels, MV-10 vertical gel electrophoresis apparatus, DC power supply, automatic micropipet, fine micropipet tips, glacial acetic acid, methanol, and distilled or deionized water.
Delivery: This kit is shipped ambient. Store lyophilized proteins at room temperature.