Vernier® Water Depth Sampler
Vernier water depth sampler
Catalog # CA470017-596
New Product
Supplier: VERNIER CANADA SCIENCE EDUCATION IN
Fournisseur #: WDS
Vernier® Water Depth Sampler
Catalog # CA470017-596
Supplier: VERNIER CANADA SCIENCE EDUCATION IN
New Product
Jump to:
Product Details & Documents
Go Deep Sea Diving With Your Water Sampler
Collect water samples from the water depth of your choice with this sampler. The sampler requires only one line to raise and lower the chamber into the waters, and will provide you with fast and accurate measurements on the sample's PH, temperature, dissolved oxygen and conductivity. Included is the sampler (1.5 L chamber) and one 50 foot nylon drop cord. Works in conjunction with the CBL, CBL2, and Labpro sensors.
Spécifications
- Désignation:Vernier water depth sampler