Water Bear
Water Bear
Supplier:   Ward's Science
Water Bear
Supplier:  Ward's Science
Certificates
Water Bear
2 Options Available Below
Product Details & Documents
Hypsibius sp. The water bear is a complex animal representing the unusual phylum of Tardigrades, which contains about 400 marine, terrestrial, and freshwater species.
  • Unusual phylum
  • Complex animals
  • Can survive in adverse conditions
These complex animals can survive extremely adverse conditions, making them ideal research subjects for studies on the aging process. Culture instructions are included. Size: up to 1 mm.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Water Bear, For 35 Students
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $23.60
    $23.60
    Each
       $23.60
    Supplier Number: CALBEAR
  • Water Bear, For 100 Students
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $47.30
    $47.30
    Each
       $47.30
    Supplier Number: -