Wash Bottle Set
Wash Bottle, Safety Labeled, 500ml Distilled Water, Wide Mouth, Blue Cap, Bag/6 Scienceware
Catalog # 470145-022
Supplier: Bel-Art
Supplier #: F11646-0620_SE
Jump to:
Product Details & Documents
For organized and convenient storage, this set of polyethylene printed wash bottles are labeled.
Packaging: Set includes six bottles labeled distilled water. Bottles are 500 ml capacity.
Specifications
- Désignation:Wash Bottle, Safety Labeled, 500ml Distilled Water, Wide Mouth, Blue Cap, Bag/6 Scienceware