Ward's® WARDSafe™ | Avantor
Ward's® WARDSafe™
Supplier:   Ward's Science
Ward's® WARDSafe™
Supplier:  Ward's Science
Certificates
Ward's® WARDSafe™
Ward's® WARDSafe™
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Specimen Holding Fluid, Dilute one part to nine parts water for use.
Shelf Life (months): 36
Storage: Red
  • Shelf Life (months): Indefinite
  • Storage: Red
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Product Family Options
Informations sur le produit
  • Désignation
    Grade
    Taille
    Chemical/Physical Specifications
    Availability
    Price
  • WARDSafe™
    Laboratory
    3.8 L
    Bottle
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (3,800ml)
    Each (3,800ml) - $134.75
    $134.75
    Each (3,800ml)
       $134.75
    Supplier Number: WW0095-3.8L
  • WARDSafe™
    Laboratory
    19 L
    Handi-Pak
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (19l)
    Each (19l) - $345.85
    $345.85
    Each (19l)
       $345.85
    Supplier Number: WW0095-19L