Wall-Mount Drying Rack
41.3 cm (16.25"), 40.6 cm (16"), 3.8 cm (1.5")
Catalog # 470020-890
Supplier: Bel-Art
Fournisseur #: F18933-0014_SE
Jump to:
Product Details & Documents
Save Valuable Bench Space
- Mounting points fit standard peg boards
- Three sizes of repositionable V-channel pegs
- Two-tier rack includes 38 pegs
Easily holds beakers, flasks, bottles, test tubes, and cylinders. A dip tray with molded-in scalloped wells also holds long, thin labware, like burettes and pipettes. Made of rugged, ABS plastic, the rack surface wipes clean easily and will not corrode.
Spécifications
- Largeur:3.8 cm (1.5")
- Hauteur:41.3 cm (16.25")
- Longueur:40.6 cm (16")