Wall Cabinets
Wall Cabinets
Certificates
Wall Cabinets
8 Options Available Below
Product Details & Documents
Constructed of solid hardwoods, solid oak and oak plywood veneers.
  • Cabinet has two adjustable shelves
  • Constructed of solid oak and oak veneers
  • Finished natural with a chemical resistant UV finish
  • Hanging wall cabinet with locking double doors
BASE CABINETS - Constructed of solid hardwood, solid oak and oak veneers. Base cabinets with choice of three styles: door cabinet, door/drawer cabinet or drawer cabinet. Cabinets can be ganged together to create a wall of base cabinets. They are finished with a chemical resistant, earth-friendly UV finish. Tops and rubber base molding sold separately. Base molding sold by the foot.
SOLID RESIN TOPS - Finish off your base cabinets with these solid epoxy resin or phenolic resin tops. These solid tops provide superior chemical resistance. Tops include 4" back splash. Available in 1" thick black.
WALL CABINETS - Constructed of solid hardwood, solid oak and oak plywood veneers. Choose from two models: Oak veneered solid doors or solid oak framed glass doors. Glass doors feature 3/16" thick safety glass. Cabinets have a lock and two adjustable shelves. They are finished with a chemical resistant, earth-friendly UV finish.
 : SEFA compliant and MAS Certified Green.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Largeur
    Profondeur
    Hauteur
    Weight
    Availability
    Price
  • Solid double door cabinet
    Oak, red
    30"
    12"
    30"
    130 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,322.00
    $1,322.00
    Each
       $1,322.00
    Supplier Number: D03-3012
  • Solid double door cabinet
    Oak, red
    36"
    12"
    30"
    140 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,406.20
    $1,406.20
    Each
       $1,406.20
    Supplier Number: D03-3612
  • Solid double door cabinet
    Oak, red
    42"
    12"
    30"
    140 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,318.50
    $1,318.50
    Each
       $1,318.50
    Supplier Number: D03-4212
  • Solid double door cabinet
    Oak, red
    48"
    12"
    30"
    150 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,430.85
    $1,430.85
    Each
       $1,430.85
    Supplier Number: D03-4812
  • Glass double door cabinet
    Oak, red
    30"
    12"
    30"
    120 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,883.85
    $1,883.85
    Each
       $1,883.85
    Supplier Number: D06-3012
  • Glass double door cabinet
    Oak, red
    36"
    12"
    30"
    130 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,989.05
    $1,989.05
    Each
       $1,989.05
    Supplier Number: D06-3612
  • Glass double door cabinet
    Oak, red
    42"
    12"
    30"
    135 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,140.10
    $2,140.10
    Each
       $2,140.10
    Supplier Number: D06-4212
  • Glass double door cabinet
    Oak, red
    48"
    12"
    30"
    140 lbs.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,277.10
    $2,277.10
    Each
       $2,277.10
    Supplier Number: D06-4812