VWR® Cylinders with Plastic Base | Avantor
VWR® Cylinders with Plastic Base
VWR® Cylinders with Plastic Base
Supplier:  VWR International
Certificates
VWR® Cylinders with Plastic Base
4 Options Available Below
Product Details & Documents
Our high-quality borosilicate cylinders meet ASTM standards.
  • Cylinders Calibrated to Contain
  • Uniform White Enamel
  • Double Graduated Metric Scale
  • Pour-Out
  • Detachable Plastic Base
They can be used with acids, salt solutions, and organic solvents.
All cylinders have a graduated metric scale and a detachable plastic base.
Packaging: One case contains twelve cylinders.
Documents
Specifications for Product Family
Spécifications
  • Catalog No:
  • 470120-662
  • 470120-664
  • 470120-666
  • 470120-668
  • Form:
  • Tall
  • Tall
  • Tall
  • Tall
Spécifications
Product Family Options
Informations sur le produit
  • Capacité
    Intervalle de graduation
    Availability
    Price
  • 10 ml
    0.1 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $153.40
    $153.40
    Case of 12
       $153.40
    Spécifications:
    More
    Specifications
    Cat. No.
    470120-662
    Form
    Tall
  • 25 ml
    0.2 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $172.05
    $172.05
    Case of 12
       $172.05
    Spécifications:
    More
    Specifications
    Cat. No.
    470120-664
    Form
    Tall
  • 50 ml
    1 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $202.40
    $202.40
    Case of 12
       $202.40
    Spécifications:
    More
    Specifications
    Cat. No.
    470120-666
    Form
    Tall
  • 100 ml
    1 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $236.35
    $236.35
    Case of 12
       $236.35
    Spécifications:
    More
    Specifications
    Cat. No.
    470120-668
    Form
    Tall