VWR® Tall Form Beaker
Supplier: VWR International
Jump to:
À propos de cet article
High Quality at a Great Value
- Borosilicate Glass Fabrication
- Graduated Markings
- Easy-to-Measure
- Beakers Meet ASTM Standards
6 Options Available Below
Product Details & Documents
High Quality at a Great Value
- Borosilicate Glass Fabrication
- Graduated Markings
- Easy-to-Measure
- Beakers Meet ASTM Standards
Documents
Certificates
Recherche par numéro de lotProduct Family Options
Informations sur le produit
- CapacitéIntervalle de graduationPlage de graduationAvailabilityPrice
- 100 ml (3.4 oz.)10 ml20 - 80 mlPack of 12$95.27Case of 48$380.12
- 200 ml (7 oz.)25 ml25 - 150 mlPack of 12$96.06Case of 48$346.04
- 300 ml25 ml25 - 250 mlPack of 12$164.30Case of 48$604.50
- 500 ml50 ml50 - 450 mlPack of 6$91.15Case of 30$448.40
- 600 ml50 ml100 - 550 mlPack of 6$129.90Case of 24$462.45
- 1000 ml50 ml50 - 950 mlPack of 6$185.20Case of 18$503.70