Biocam Virus Chart Set
Catalog # 470023-544
Supplier: BioCam Communications
Specifications
- Width:45.7 cm
- Description:Virus Chart Set
- Length:61 cm
- Cat. no.:470023-544
- Finish:Laminated
- Supplier No.:SPECIAL
Specifications
About this item
Comprehensive Overview of Structure and Replication
- Colorful illustrations and descriptive text
These charts present useful information for your virology studies. The Virus I chart depicts well-known examples of icosahedral, helical, enveloped, and complex virus structures; Virus II chart covers replication, with detail on Influenza life cycle; Virus III presents information specific to bacteriophages, contrasting lytic and lysogenic cycles. Set of three charts.