Vernier® pH Sensors
Vernier® pH Sensors
Vernier® pH Sensors
New Product
Vernier® pH Sensors
Vernier® pH Sensors
General-purpose pH Sensors can be used across disciplines, with applications in chemistry, biology, middle school science, and environmental science.
  • Automated Data Collection
  • Graphing
  • Data Analysis
  • For Titrations, Studies of Household Acids and Bases, and More
  • Ideal for Use in Multiple Disciplines
Advantages of these sensors include automated data collection, graphing, and data analysis.
Typical activities using pH Sensors include acid-base titrations, studies of household acids and bases, monitoring pH change during chemical reactions or in an aquarium as a result of photosynthesis, investigations of acid rain and buffering, and analysis of water quality in streams and lakes.
  • Catalog No:
  • CA470039-294
  • CA470219-320
  • Response time:
  • 90% of final reading in 1 second
  • Accuracy:
  • ±0.2 pH units
  • Operating temperature range:
  • 5...80°C
Product Family Options
  • Désignation
    Electrolyte
  • Vernier pH sensor (BTA)
    Ag/AgCl Gel
    Each
    Each - $189.09
    $189.09
    Each
       $189.09
    Minimum order quantity is 1
     : PH-BTA
    New Product
    Specifications
    Cat. No.
    CA470039-294
    Response time
    90% of final reading in 1 second
    Accuracy
    ±0.2 pH units
    Operating temperature range
    5...80°C
    New Product
  • Vernier pH storage solution, 500 ml
    Each
    Each - $45.09
    $45.09
    Each
       $45.09
     : PH-SS
    New Product
    Specifications
    Cat. No.
    CA470219-320
    Response time
    Accuracy
    Operating temperature range