Vernier® CO₂ Gas Sensor
Vernier CO₂ gas sensor (BTA)
CA470039-278
New Product
: CO2-BTA
The CO₂ gas sensor measures gaseous carbon dioxide.
- Wired connection with BTA plug
The CO₂ gas sensor measures gaseous carbon dioxide in two ranges –0 to 10,000 ppm and 0 to 100,000 ppm. With this sensor, you can easily monitor changes in CO₂ levels occurring in respiration of organisms ranging from peas to humans. A 250 ml respiration chamber with probe attachment is included for running controlled experiments with small plants and animals.
: LabQuest® (2, 3, Mini) interface is required but not included.
- Désignation:Vernier CO₂ gas sensor (BTA)
- Cat. No.:CA470039-278
- Range:Low: 0 - 10000 ppm; High: 0 - 100000 ppm