Vermiculite | Avantor
Vermiculite
Vermiculite, 4 qts.
Catalog # 470021-148
Supplier:   Avantor
Vermiculite
Catalog # 470021-148
Supplier:  Avantor
Vermiculite
Vermiculite
Vermiculite, 4 qts.
Price
16,20 $
Log in pour la disponibilité
Pack of 1
Product Details & Documents
Light, porous mica medium retains moisture well, making it excellent for starting seeds or as a soil additive.
Spécifications
  • Désignation:
    Vermiculite, 4 qts.