Variable Speed Electrostatic Generator
À propos de cet article
This generator can perform all electrostatic experiments on a spectacular scale.
- Rugged Construction
- Troubleshooting Guide Included
- Generator Produces 250,000 to 400,000 Volts
- Polished, Smudge-Resistant Finish
- Extra Belt Included
This variable speed Van de Graaff-type generator produces 250,000 to 400,000 volts. It's built to give maintenance-free operation for many years. The sphere is 25 cm in diameter with a polished, smudge-resistant finish. The column between the base and sphere is made of clear Lucite.
The overall height of the unit is 76 cm. The variable speed motor is controlled by a knob and can be used to vary the rate of charge buildup. One extra belt is included with the unit.
Informations sur les accessoires: Replacement belts, 3.8 x 67.3 cm circumference, are available separately.
2 Options Available Below
Product Details & Documents
This generator can perform all electrostatic experiments on a spectacular scale.
- Rugged Construction
- Troubleshooting Guide Included
- Generator Produces 250,000 to 400,000 Volts
- Polished, Smudge-Resistant Finish
- Extra Belt Included
This variable speed Van de Graaff-type generator produces 250,000 to 400,000 volts. It's built to give maintenance-free operation for many years. The sphere is 25 cm in diameter with a polished, smudge-resistant finish. The column between the base and sphere is made of clear Lucite.
The overall height of the unit is 76 cm. The variable speed motor is controlled by a knob and can be used to vary the rate of charge buildup. One extra belt is included with the unit.
Informations sur les accessoires: Replacement belts, 3.8 x 67.3 cm circumference, are available separately.
Specifications for Product Family
Spécifications
- Catalog No:
- 470005-526
- 470005-528
- Output:
- 350 kV
- −
Spécifications
Product Family Options
Informations sur le produit
- DésignationDiameterTailleAvailabilityPrice
- Supplier Number: N-100ESpécifications:
More
Specifications
Cat. No.470005-526Output350 kV - Supplier Number: RP-601Spécifications:
More
Specifications
Cat. No.470005-528Output−