Vacuum Desiccators | Avantor
Vacuum Desiccators
Vacuum Desiccators
Certificates
Vacuum Desiccators
Vacuum Desiccators
2 Options Available Below
Product Details & Documents
Compact design for easy use.
  • Autoclavable
  • Clear design
These autoclavable desiccators are made of polypropylene and polycarbonate, and can hold a vacuum up to 740 mm Hg for 24 hours without any greasing. Autoclavable at 121 °C for 15 minutes. The top dome, molded in rigid and transparent PC, gives a crystal-clear view of the sample. The knurled knob on the top provides easy handling of the dome.
The lower part is made of PP and includes a perforated PP desiccator plate. An internal groove on the flange holds the silicon rubber O-ring. The PP stopcock accepts 1/4" I.D. tubing and has a self-lubricating PTFE plug. Use the three-way plug to create a vacuum, release a vacuum, or for shut-off.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Vacuum Desiccators, 6" Dia.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $174.25
    $174.25
    Each
       $174.25
    Supplier Number: 55204
  • Vacuum Desiccators, 10" Dia.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $282.05
    $282.05
    Each
       $282.05
    Supplier Number: 55206