Unmounted Mirrors
Unmounted Mirrors
Unmounted Mirrors
Certificates
Unmounted Mirrors
Unmounted Mirrors
3 Options Available Below
Product Details & Documents
Converging and diverging mirrors for optical experiments.
  • Optically true glass
  • Scratch-resistant copper back silvering
An excellent selection of spherical, silvered glass mirrors for demonstrations and experiments in optics.
Product Family Options
Product Information
  • Désignation
    Focal Length
    Diameter
    Availability
    Price
  • Convex mirror
    50 mm
    50 mm
    Log in for availability
    Log in for availability
    Each
    Each - $12.65
    $12.65
    Each
       $12.65
    Supplier Number: MCV502
  • Convex mirror
    76 mm
    75 mm
    Log in for availability
    Log in for availability
    Each
    Each - $11.05
    $11.05
    Each
       $11.05
    Supplier Number: MCV703
  • Concave mirror
    50 mm
    50 mm
    Log in for availability
    Log in for availability
    Each
    Each - $11.20
    $11.20
    Each
       $11.20
    Supplier Number: MCC502