Fischer Technical Universal Vacuum Plate
Fischer Technical Universal Vacuum Plate
Plate, universal vacuum
  470006-052
Supplier #:  GA-001
Fischer Technical Universal Vacuum Plate
  470006-052
Fischer Technical Universal Vacuum Plate
Fischer Technical Universal Vacuum Plate
Plate, universal vacuum
$697.45
Each
This nickel coated plate uses a rubber pad to achieve air tight contact with a bell jar. It features a stopcock and center hole with safety screw plug and a rubber gasket.
To create the vacuum, air is removed via a barbed hose connector that accommodates tubing of 6.4 mm (¼") int.Ø. Measures diameter 27 cm with a 6 mm diameter center hole.
  • Désignation:
    Plate, universal vacuum