Universal Precaution Compliance Kit
Universal Precaution Compliance Kit
Catalog # 470018-446
Supplier: MASUNE INC.
Fournisseur #: 19151M
Jump to:
Product Details & Documents
Protect yourself, and clean up your lab, with one handy combination kit. Items for protection against pathogen spills, as well as equipment for safe removal and disposal of spilled materials, all of which meet OSHA, CDC, and state requirements, are provided. The kit includes a safety shield with mask, disposable apron, medical gloves, Red Z pouch, scoop with scraper, germicidal disinfectant surface wipe, biohazard waste bag, antimicrobial hand wipe, and identification tag.
Spécifications
- Désignation:Universal Precaution Compliance Kit