Ulva, Thallus Slide
Ulva, Thallus, Whole mount, Fast Green and Mallory's
Catalog # 470180-990
Supplier: Avantor
Jump to:
Product Details & Documents
Sea Lettuce
- Identifying features clearly distinguishable
- Rigorous quality control standards
Edible vegetation in the genus Ulva.
Spécifications
- Désignation:Ulva, Thallus
- Stain:Fast Green and Mallory's
- Orientation:Whole mount