ContactUsLinkComponent
Ultrasonic Cleaner
  470105-414
undefined
Ultrasonic Cleaner
  470105-414
 :  311
  • Capacity:
    3 L
  • Cat. no.:
    470105-414
  • W×D×H:
    14×10×24 cm
  • Electrical:
    120 VAC, 60 Hz, 95 W
  • Supplier No.:
    311
Clean the Dirtiest or Most Delicate Laboratory Equipment with Ease.
  • Features include a large tank, on/off switch, and convenient timer
  • Tank features solid-state circuitry
Ultrasonic cleaning systems provide powerful surges of cavitation energy to reach interior areas and the tiniest grooves that hand cleaning can’t touch. Features include a large tank, on/off switch, and convenient timer. Tank is made of durable stainless steel/vinyl-clad metal. Complete with stainless steel lid. Basket not included.