Ward's® Ulothrix | Avantor
Ward's® Ulothrix
Supplier:   Ward's Science
Ward's® Ulothrix
Supplier:  Ward's Science
Certificates
Ward's® Ulothrix
Ward's® Ulothrix
2 Options Available Below
Product Details & Documents
Freshwater; unbranched cells with parietal chloroplasts; produces zoospores.
Live Cultures are supplied with media in 16 x 125 mm tubes, 2 oz. jars, or 8 oz. jars. Fast delivery is available directly from our laboratory. Please specify a delivery date.
Each culture shipment is prepared in our laboratories and includes the "Working with Algae & Cyanobacteria" manual which contains detailed instructions for subculturing and use.
Canadian Customers:
CFIA and PHAC regulations require customers to submit a completed inspection checklist and signed statement of facilities compliance before receiving certain cultures. A customer service representative will contact you if documentation is required.
Product Family Options
Informations sur le produit
  • Désignation
    Media
    No. of Students
    Packaging
    Availability
    Price
  • Ulothrix, for 120 Students
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $293.93
    $293.93
    Each
       $293.93
    Supplier Number: -
  • Ulothrix (Small)
    Basic culture solution
    30
    Tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $97.93
    $97.93
    Each
       $97.93
    Supplier Number: ULOT30/35