Preserved Turtles
Preserved Turtles
Supplier:   Avantor
Preserved Turtles
Supplier:  Avantor
Certificates
Preserved Turtles
1 Options Available Below
Product Details & Documents
Formalin preserved turtles available in pails.
  • Reptile anatomy
  • General dissection
  • High quality preservation
We offer Pseudemys sp. turtles for study specimens that can be used to teach comparative anatomy and the similarities and differences between reptiles, amphibians, and mammals. Students can explore the structures of the shell, musculature, and internal organs of the study animal. The carapace is pre-cut for ease of dissection. We offer plain preserved, single, double, or triple injected turtles for your teaching or research needs.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Injection Type
    Packaging
    Availability
    Price
  • Formalin Preserved Turtles
    4" to 6"
    Uninjected
    Pail of 10
    Discontinued
    Supplier Number: -
  • Formalin Preserved Turtles
    6 to 8"
    Plain preserved
    Pail of 1
    Discontinued
    Supplier Number: 137.2126.1
  • Formalin Preserved Turtles
    6 to 8"
    Plain preserved
    Pail of 10
    Discontinued
    Supplier Number: 137.2126.10
  • Formalin Preserved Turtles
    8" to 10"
    Uninjected
    Pail of 5
    Discontinued
    Supplier Number: -
  • Formalin Preserved Turtles
    8 to 10"
    Plain preserved
    Pail of 1
    Discontinued
    Supplier Number: 137.2102.1
  • Formalin Preserved Turtles
    8 to 10"
    Plain preserved
    Pail of 10
    Discontinued
    Supplier Number: -
  • Formalin Preserved Turtles
    8 to 10"
    Single injected
    Pail of 10
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $838.00
    $838.00
    Pack of 10
       $838.00
    Supplier Number: -
  • Formalin Preserved Turtles
    8 to 10"
    Double injected
    Pail of 1
    Discontinued
    Supplier Number: 137.2114.1
  • Formalin Preserved Turtles
    8 to 10"
    Double injected
    Pail of 10
    Discontinued
    Supplier Number: -
  • Formalin Preserved Turtles
    8 to 10"
    Triple injected
    Pail of 1
    Discontinued
    Supplier Number: -
  • Formalin Preserved Turtles
    8 to 10"
    Triple injected
    Pail of 10
    Discontinued
    Supplier Number: -
  • Formalin Preserved Turtles
    8" to 10"
    Double injected
    Pail of 5
    Discontinued
    Supplier Number: -
Accessories
Informations sur le produit
  • Catalog #
    Grade
    Size
    Availability
    Price
  • WARDSafe™
    WARDSafe™
    470233-546
    Laboratory
    19 L
    Log in pour la disponibilité
    Each (19l) - $345.85