Turbidity | Avantor
Turbidity
Supplier:   LaMotte
Turbidity
Supplier:  LaMotte
Certificates
Turbidity
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Pre-calibrated test tubes and labware give fast, accurate results in the classroom or in the lab. 
  • Range: Five to 200 JTU
  • Material for 50 Tests
  • Compact, Waterproof Carrying Case Contains Kit
  • Instructions Included
  • Pre-calibrated Test Tubes
A compact, waterproof carrying case contains reagents, labware, and step-by-step instructions.
The tests have a range of 5 to 200 JTU.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Turbidity Refill Kit
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $32.60
    $32.60
    Pack of 1
       $32.60
    Supplier Number: 7520-H
  • LaMotte® Water Test Kit, Turbidity
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 1
    Kit of 1 - $177.75
    $177.75
    Kit of 1
       $177.75
    Supplier Number: 7519-01