T-Shaped Flint Glass Connecting Tubes
470235-772
- Ext.Ø:5/16"
- Ø int.:1/4"
- Description:T-Shaped Flint Glass Connecting Tubes, Arm Length: 1 1/2"
- Material:Flint glass
- Cat. no.:470235-772
For joining two rubber tubes to a common tube at right angles. Safer connections are provided as the ends have constrictions to hold rubber tubing securely.
- Made from quality flint glass