Trundle Wheel
Trundle wheel, 1 m circumference, 60 cm (handle)
Catalog # 470110-724
Supplier: SI Manufacturing
Fournisseur #: 45550
Jump to:
Product Details & Documents
A Fast and Simple Way to Measure Long Distances.
- Wheel circumference is 1 m
- Produces audible click with every revolution for tracking purposes
- 60 cm long handle
Spécifications
- Désignation:Trundle wheel, 1 m circumference
- Longueur:60 cm (handle)