Tris-Borate-EDTA Solution
Tris-Borate-EDTA Solution
Supplier:   Ward's Science
Tris-Borate-EDTA Solution
Supplier:  Ward's Science
Certificates
Tris-Borate-EDTA Solution
Tris-Borate-EDTA Solution
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Tris-Borate-EDTA Solution is premixed.
  • Powder form
  • For use with Borate gels only
  • Premixed
It is in powder form.
The solution is for use with borate gels only.
Certifications: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Caution: Note: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Product Family Options
Product Information
  • Désignation
    Grade
    Taille
    Chemical/Physical Specifications
    Availability
    Price
  • Tris-Borate-EDTA Running Buffer
    Laboratory
    Makes 6 L
    1X Solution
    Log in for availability
    Log in for availability
    Each
    Each - $71.35
    $71.35
    Each
       $71.35
    Minimum order quantity is 1
    Supplier Number: TT0283-102G
  • Tris-Borate-EDTA Running Buffer
    Laboratory
    Makes 2.5 L
    1X Solution
    Log in for availability
    Log in for availability
    Each
    Each - $51.25
    $51.25
    Each
       $51.25
    Supplier Number: TT0280-500
  • Tris-Borate-EDTA Running Buffer
    Laboratory
    Makes 2.5 L
    1X Solution
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $42.75
    $42.75
    Each (500ml)
       $42.75
    Supplier Number: TT0280-500ML