Tree Calipers
Tree Calipers
Catalog # 470021-026
Supplier: Peco Sales
Supplier #: 59722
Jump to:
Product Details & Documents
Easily Take Accurate Measurements in the Field
Made of aluminum, this lightweight tree caliper is a valuable forestry tool designed for ease of use. Measure the diameter of tree trunks and other large objects up to 24 inches with the precision of 1/10 inch graduations marked on both sides. Size: 34" x 16".
Specifications
- Désignation:Tree Calipers