Transitional Epithelium Slide
Transitional Epithelium Slide, Section, Hematoxylin and Eosin
Catalog # 470182-618
Supplier: Avantor
Jump to:
Product Details & Documents
Multilayered epithelium
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Stratified cuboidal epithelium from excretory system.
Spécifications
- Désignation:Transitional Epithelium Slide
- Stain:Hematoxylin and Eosin
- Orientation:Section