Toxocara canis, Eggs Slide
Toxocara canis, Eggs, Whole mount, Giemsa
Catalog # 470182-184
Supplier: Avantor
Jump to:
Product Details & Documents
A Common Parasite in Canids
- Identifying features clearly distinguishable
- Rigorous quality control standards
Ingestion of these embryonated eggs is responsible for visceral larva migrans in humans. Unstained. The Toxocara canis slide is double cover mounted, and therefore is best viewed using 4 and 10x microscope objectives.
Spécifications
- Désignation:Toxocara canis, Eggs
- Stain:Giemsa
- Orientation:Whole mount