Toolmark ID Poster
Toolmark ID Poster
Catalog # 470023-272
Supplier: PRECISION FORENSICS TESTING
Supplier #: PT-01
Jump to:
Product Details & Documents
Realistic Pictures Help in Identification
- Size: 24" x 36"
Many different tools and their respective marks are highlighted on this explanatory poster. The high quality pictures clearly demonstrate the essential aspects of toolmark identification. Size: 24" x 36".
Specifications
- Désignation:Toolmark ID Poster