ContactUsLinkComponent
Screwdriver and Wrench Set
Screwdriver and Wrench Set
Catalog # 470123-726
Supplier:  Moody Tools
undefined
Screwdriver and Wrench Set
Catalog # 470123-726
Supplier:  Moody Tools
Supplier Number:  58-0154
Specifications
  • Description:
    Screwdriver and Wrench Set
  • Cat. no.:
    470123-726
  • Supplier No.:
    58-0154
Specifications
About this item
This set is designed for minor repair and adjustment of laboratory apparatus.
Ordering information: The set consists of 27 interchangeable tips, a chuck type, and a knurled handle with a swivel head. It includes eight open-end and five socket wrenches in sizes 5/16, 3/32, 7/64, 1/8, and 5/32 inch; six regular screwdriver tips, sizes 0.025, 0.040, 0.055, 0.070, 0.080, and 0.100 inch; three Allen-type hex keys numbers 4, 6, and 8; and six hex driver blades with number 0 and number 1 Phillips-type screwdriver tips.