T-Nesting Support Bases
T-Nesting Support Bases, T Size L x W, in.: 7-1/8 x 8-3/4, Thread Size: 3/8 - 16, Rod Dia., in.: 1/2
Catalog # 470123-486
Supplier: Wards
Fournisseur #: WLS78355-10C
Jump to:
Product Details & Documents
Specially designed to provide excellent stability, these bases are shaped with legs that slide freely under one another for compact storage or set up.
Spécifications
- Désignation:T-Nesting Support Bases, T Size L x W, in.: 7-1/8 x 8-3/4, Thread Size: 3/8 - 16, Rod Dia., in.: 1/2