Tissue Forceps With Teeth
Tissue Forceps With Teeth
Tissue Forceps With Teeth
Tissue Forceps With Teeth
A true no slip hold.
  • Interlocking teeth prevent slipping
  • Ridged handles for a confident hold
  • Stainless construction prevents chipping
For general dissection, these 1 × 2 teeth firmly grasp tissues, while the serrated handles provide nonslip control. Gillie's type with straight blunt tips.
 : These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
  • Désignation
    Matière
    Longueur
    Tip Style
  • Tissue Forceps
    Stainless steel
    115 mm (4.5")
    1 x 2 Tooth
    Each
    Each - $12.15
    $12.15
    Each
       $12.15
     : FOBL45
  • Tissue Forceps
    Stainless steel
    110 mm (4.5")
    2 x 3 Tooth
    Each
    Each - $12.55
    $12.55
    Each
       $12.55
     : 141015
  • Tissue Forceps
    Stainless steel
    152 mm (6")
    1 x 2 Tooth
    Each
    Each - $12.30
    $12.30
    Each
       $12.30
     : 141017