Thioglycollate Medium
Thioglycollate Medium
Prepared Thioglycollate Medium Tubes
Catalog # 470180-662
Supplier:   Avantor
Thioglycollate Medium
Catalog # 470180-662
Supplier:  Avantor
CAS Number:  
Thioglycollate Medium
Thioglycollate Medium
Prepared Thioglycollate Medium Tubes
CAS Number:  
Price
$66.30
Log in for availability
Pack of 12
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Without indicator. For cultivation of anaerobic species and Clostridium. Suitable for fermentation studies where no indicator is required.
Specifications
Specifications
  • Désignation:
    Prepared Thioglycollate Medium Tubes
Specifications