Thin Stem Pipettes
Thin Stem Pipettes
Supplier:   Kemtec
Thin Stem Pipettes
Supplier:  Kemtec
Certificates
Thin Stem Pipettes
Thin Stem Pipettes
2 Options Available Below
Product Details & Documents
Versatile pipettes provide consistent drop size.
  • Chemically inert and non-toxic
  • Can be stretched to form micropipettes and other micro lab ware
These pipettes serve as ideal micro-reagent vials that can be heat-sealed and frozen.
Product Family Options
Informations sur le produit
  • Capacité
    Drops per mL
    Availability
    Price
  • 4 ml
    25
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $59.70
    $59.70
    Pack of 100
       $59.70
    Supplier Number: 4-424C
  • 4 ml
    25
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 500
    Pack of 500 - $210.45
    $210.45
    Pack of 500
       $210.45
    Supplier Number: 4-424L