ContactUsLinkComponent
Thermostat
  470005-892
 :  Wards
undefined
Thermostat
  470005-892
 :  Wards
 :  PH0403
  • Description:
    Thermostat, Working Model, 3 Amp
  • Cat. no.:
    470005-892
  • Supplier No.:
    PH0403
This working model thermostat will demonstrate how differential expansion is used to control temperature. A bimetallic strip is used to open or close an electric circuit when temperature changes. The contacts will carry about three amperes. The unit can be operated with a 6 V lantern battery, low voltage power supply, or a blow dryer. Mounted on a metal base; approximately 15 cm high.