Thermometer/Hygrometer
Thermometer/Hygrometer
470012-294
: 497J
Built to Last for Years
With its rustproof parts and shatterproof crystal, this combination thermometer/hygrometer will stand up to years of use. It measures temperature and humidity in any area, indoors or out and the bright graphics are easily visible even from a distance. Relative humidity: 20–90%; temperature range: -50°F to +130°F (-46°C to +54°C). Size: 12" dia.
- Désignation:Thermometer/Hygrometer