The Rock Cycle Study Kits | Avantor
The Rock Cycle Study Kits
The Rock Cycle Study Kits
Certificates
The Rock Cycle Study Kits
The Rock Cycle Study Kits
2 Options Available Below
Product Details & Documents
What causes rocks to change over time? This hands-on kit lets students use igneous, sedimentary and metamorphic specimens to examine processes such as weathering, lithification, metamorphism and magmatic assimilation.
Kit includes:
• Samples and study charts for 30 student groups
• Large specimens for easy examination
• Chart with mounted specimens
• Detailed study guide with physical properties and descriptions of more than 50 rocks, minerals, and ores.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Chart, The rock cycle, 60×39 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $190.80
    $190.80
    Each
       $190.80
    Supplier Number: 2521
  • Kit, The rock cycle, complete/60 students in pairs
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 1
    Kit of 1 - $379.45
    $379.45
    Kit of 1
       $379.45
    Supplier Number: 2721