LaMotte® Salinity Test Kit
This kit test for salinity using the Direct Reading Titrator method.
- Aligned with GLOBE protocols
This kit test for salinity using the Direct Reading Titrator method.
- Aligned with GLOBE protocols
- Catalog No:
- 470203-126
- 470021-206
- Increments:
- 0.1 ppt salinity
- Sensitivity:
- 0.4 ppt salinity
Product Family Options
- DésignationMéthode chimiqueRangeNo. of Tests
- : 7459-02
Specifications
Cat. No.470203-126Increments0.1 ppt salinitySensitivity0.4 ppt salinity - : R-7459-01
Specifications
Cat. No.470021-206IncrementsSensitivity