The Chick Embryo Microslide
The Chick Embryo Microslide
The Chick Embryo Microslide
Certificates
The Chick Embryo Microslide
The Chick Embryo Microslide
2 Options Available Below
Product Details & Documents
Explore chick development on the microscopic level.
  • Arrows help students locate important features
  • Mounted in a clear plastic holder that protects it on both sides
Microslides consist of a strip of film with photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own microslides. A detailed text folder is included. Viewers are available separately.
 : Set includes 13 hours (wm, st, 1X), 18 hours (wm, st, 1X), 21 hours (wm, st, 1X), 28 hours (wm, st, 1X), 38 hours (wm, st, 1X), 48 hours(wm, st, 1X), 56 hours (wm, st,1X), and 96 hours (wm, st, 1X).
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • The chick embryo microslide
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $19.60
    $19.60
    Pack of 1
       $19.60
    Supplier Number: T-17
  • The chick embryo microslides, set of 15
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 15
    Kit of 15 - $292.00
    $292.00
    Kit of 15
       $292.00
    Supplier Number: T-17-15B