Testis, Spermatogenesis, Mammal Slide
Testis, Spermatogenesis, Mammal Slide, Section, Iron Hematoxylin
Catalog # 470177-626
Supplier: Avantor
Jump to:
Product Details & Documents
Male reproductive components
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Shows capsule, seminiferous tubules, spermatogenic cells, sertoli, and Leydig’s cells.
Spécifications
- Désignation:Testis, Spermatogenesis, Mammal Slide
- Stain:Iron Hematoxylin
- Orientation:Section