Termite, Worker Slide
Termite, Worker, Whole mount, Unstained
Catalog # 470181-612
Supplier: Avantor
Jump to:
Product Details & Documents
Illustrates Body Structures
- Identifying features clearly distinguishable
- Rigorous quality control standards
Worker termite demonstrating general body structures and biting mouthparts. Highly cooperative social structure.
Spécifications
- Désignation:Termite, Worker
- Stain:Unstained
- Orientation:Whole mount