Preserved Lampshell | Avantor
Preserved Lampshell
Formalin Preserved Lampshell, 3/4", Uninjected, Jar of 10
Catalog # 470001-234
Supplier:   Avantor
Preserved Lampshell
Catalog # 470001-234
Supplier:  Avantor
Preserved Lampshell
Preserved Lampshell
Formalin Preserved Lampshell, 3/4", Uninjected, Jar of 10
Price
$153.99
Log in pour la disponibilité
Pack of 10
Product Details & Documents
Formalin preserved lampshell. Available in jars.
  • Invertebrate anatomy
  • High quality preservation
Either Terrbratalia sp. or Terebratulina sp.This unique brachiopod genus is native to the Pacific coast. Size: 3/4".
Spécifications
  • Désignation:
    Formalin Preserved Lampshell
  • Taille:
    3/4"
  • Injection Type:
    Uninjected
  • Packaging:
    Jar of 10
Accessories
Informations sur le produit
  • Catalog #
    Grade
    Size
    Availability
    Price
  • WARDSafe™
    WARDSafe™
    470233-546
    Laboratory
    19 L
    Log in pour la disponibilité
    Each (19l) - $345.85