Ward's® Tenacity Collection
Tenacity Collection
Catalog # 470025-246
Supplier: Avantor
Jump to:
Product Details & Documents
Five Types of Cohesiveness
Different types of tenacity are shown in five popular minerals. The set is housed in a compartmented collection box, and an identification list is included.Contents:5 numbered samples, 1" x 1"Minerals:Brittle: sulfurSectile: seleniteMalleable: copperFlexible: vermiculiteElastic: mica
Specifications
- Désignation:Tenacity Collection