Dissecting Needles
Dissecting Needles
Supplier:   Dunrite Instruments
Dissecting Needles
Certificates
Dissecting Needles
Dissecting Needles
2 Options Available Below
Product Details & Documents
A must for any dissection.
  • Use to tease muscle or tissue apart
  • Selection of wood, metal, or plastic handles
  • Straight or bent tips
Choose a size to suit your personal preference. Straight needles are used for general work, while bent tips make tracing vessels and nerves easy. Available in all stainless steel, or stainless needles in plastic or wood handles.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Product Family Options
Product Information
  • Désignation
    Matière
    Longueur
    Tip Style
    Availability
    Price
  • Teasing Needle
    Plastic
    146 mm (5.75")
    Straight
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $3.80
    $3.80
    Log in for availability
    Pack of 12
    Pack of 12 - $44.85
    $44.85
    Each
       $3.80
    Pack of 12
       $44.85
    Supplier Number: NEDPL1-PK/12
  • Straight Needle
    Wood
    140 mm (5.5")
    Straight
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $4.40
    $4.40
    Log in for availability
    Case of 6
    Case of 6 - $16.10
    $16.10
    Each
       $4.40
    Case of 6
       $16.10
    Supplier Number: WLS26805