Teaching Sphygmomanometer
Sphygmomanometer, Teaching Set
  470105-208
 :   Avantor
Teaching Sphygmomanometer
  470105-208
 :  Avantor
Teaching Sphygmomanometer
Teaching Sphygmomanometer
Sphygmomanometer, Teaching Set
$127.35
Each Pallet of 48
The Teaching Sphygmomanometer is an excellent instructional tool.
  • Aneroid Sphygmomanometer
  • Dual-Earpiece Stethoscope
  • For Simultaneous Listening
It includes our aneroid sphygmomanometer plus our dual-earpiece stethoscope.
The unit allows a teacher and student or two students to listen simultaneously.
  • Désignation:
    Sphygmomanometer, Teaching Set