Preserved Taenia sp.
Catalog # 470001-814
Supplier: Avantor
Specifications
- Description:Formalin Preserved Taenia sp., Adult
- Size:Varies
- Cat. no.:470001-814
- Packaging:Jar of 1
- Supplier No.:WARD470001-814
Specifications
About this item
Formalin preserved Taenia. Available in jars or vials.
- Invertebrate anatomy
- High quality preservation
Cat/dog tapeworm. Scolex and proglottids in various stages of maturity.