Taenia pisiformis, Eggs Slide
Catalog # 470182-114
Supplier: Avantor
Specifications
- Description:Taenia pisiformis, Eggs
- Cat. no.:470182-114
- Stain:Unstained
- Supplier No.:WARD470182-114
- Orientation:Whole mount
Specifications
About this item
Endoparasitic Tapeworm
- Identifying features clearly distinguishable
- Rigorous quality control standards
Egg of Taenia pisiformis. The so-called “shell” is actually an embryonic membrane, the embryophore. The embryo is within. Unstained.